Assay Details
Target Gene Details
Entrez Gene ID: | 2825 |
Gene Name: | G protein-coupled receptor 1 |
Gene Aliases: |
- |
Location: |
Chr.2:206175316-206218047 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR1 | NM_001098199.1 | 3 | 1007 | NP_001091669.1 |
| NM_001261452.1 | 4 | 1258 | NP_001248381.1 | |
| NM_001261453.1 | 4 | 1262 | NP_001248382.1 | |
| NM_001261454.1 | 3 | 853 | NP_001248383.1 | |
| NM_001261455.1 | 3 | 849 | NP_001248384.1 | |
| NM_005279.3 | 3 | 1011 | NP_005270.2 | |
| XM_005246471.2 | 3 | 773 | XP_005246528.1 | |
| XM_017003832.1 | 3 | 1710 | XP_016859321.1 | |
| AK075130.1 | 2 | 718 | ||
| AK290976.1 | 3 | 925 | ||
| AK312917.1 | 4 | 1114 | ||
| AL550780.3 | 2 | 706 | ||
| BC067833.1 | 3 | 876 | AAH67833.1 | |
| BX094581.1 | 1 | 244 | ||
| EF577403.1 | 1 | 625 |
More Information
Set Membership: |
|

Genomic Map