Assay Details
Target Gene Details
Entrez Gene ID: | 29074 |
Gene Name: | mitochondrial ribosomal protein L18 |
Gene Aliases: |
HSPC071, L18mt, MRP-L18 |
Location: |
Chr.6:159789812-159798429 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL18 | NM_001318817.1 | NP_001305746.1 | ||
| NM_014161.4 | NP_054880.2 | |||
| NR_134860.1 | ||||
| NR_134861.1 | ||||
| NR_134862.1 | ||||
| NR_134863.1 | ||||
| NR_134864.1 | ||||
| NR_134865.1 | ||||
| NR_134866.1 | ||||
| AF161556.1 | AAF29043.1 | |||
| AK311391.1 | ||||
| AL136633.1 | CAB66568.1 | |||
| AV756740.1 | ||||
| BC001623.1 | AAH01623.1 | |||
| BF797811.1 | ||||
| BG548185.1 | ||||
| BM924328.1 | ||||
| BU601065.1 | ||||
| BU932963.1 | ||||
| CA777760.1 | ||||
| CB529016.1 | ||||
| CD675657.1 | ||||
| CR533491.1 | CAG38522.1 |
More Information
Set Membership: |
|

Genomic Map