Assay Details
Target Gene Details
Entrez Gene ID: | 3300 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B2 |
Gene Aliases: |
CMT2T, DSMA5, HSJ-1, HSJ1, HSPF3 |
Location: |
Chr.2:219279318-219286900 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB2 | NM_001039550.1 | NP_001034639.1 | ||
| NM_006736.5 | NP_006727.2 | |||
| AK289626.1 | ||||
| AK292761.1 | ||||
| AK293888.1 | ||||
| AK312723.1 | ||||
| AL547521.3 | ||||
| BC047056.1 | AAH47056.1 | |||
| BT007088.1 | AAP35751.1 | |||
| S37374.1 | AAA09035.1 | |||
| S37375.1 | AAA09034.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584501 | Chr.2:219260522 - 219323181 on Build GRCh38 | Loss |
|
| nsv527936 | Chr.2:219270486 - 219328674 on Build GRCh38 | Loss |
|
| nsv834543 | Chr.2:219226398 - 219416430 on Build GRCh38 | Loss |
|
| nsv508885 | Chr.2:219231225 - 219333602 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map