Assay Details
Target Gene Details
Entrez Gene ID: | 25934 |
Gene Name: | nipsnap homolog 3A |
Gene Aliases: |
HSPC299, NIPSNAP4, TASSC |
Location: |
Chr.9:104747688-104760122 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NIPSNAP3A | NM_015469.1 | 6 | 1140 | NP_056284.1 |
| AF161417.1 | 6 | 1121 | AAF28977.1 | |
| AK024015.1 | 6 | 1101 | BAB14777.1 | |
| AL117557.1 | 6 | 1084 | CAB55992.2 | |
| BC005935.1 | 6 | 1140 | AAH05935.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891706 | Chr.9:104480016 - 105304573 on Build GRCh38 | Gain |
|
| nsv521783 | Chr.9:104757168 - 104770036 on Build GRCh38 | Loss |
|
| esv3891705 | Chr.9:104391828 - 106660921 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map