Assay Details
Target Gene Details
Entrez Gene ID: | 152273 |
Gene Name: | FYVE, RhoGEF and PH domain containing 5 |
Gene Aliases: |
ZFYVE23 |
Location: |
Chr.3:14818962-14934565 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGD5 | NM_001320276.1 | 1 | 1056 | NP_001307205.1 |
| NM_152536.3 | 1 | 1056 | NP_689749.3 | |
| XM_011533422.1 | 1 | 1056 | XP_011531724.1 | |
| XM_011533423.1 | 1 | 1056 | XP_011531725.1 | |
| AK131078.1 | 1 | 720 | BAC85128.1 | |
| BC132814.1 | 1 | 694 | ||
| BC132816.1 | 1 | 694 | ||
| BC144300.1 | 1 | 694 | ||
| BX640820.1 | CAE45896.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2724945 | Chr.3:13931758 - 14820330 on Build GRCh38 | Deletion |
|
| nsv589778 | Chr.3:14765868 - 14823860 on Build GRCh38 | Loss |
|
| esv3893664 | Chr.3:14808160 - 14845522 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map