Assay Details
Target Gene Details
Entrez Gene ID: | 55335 |
Gene Name: | nipsnap homolog 3B |
Gene Aliases: |
FP944, NIPSNAP3, SNAP1 |
Location: |
Chr.9:104763741-104777764 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NIPSNAP3B | NM_018376.3 | NP_060846.2 | ||
| NR_130759.1 | ||||
| NR_130760.1 | ||||
| XM_011518839.1 | XP_011517141.1 | |||
| XM_017014889.1 | XP_016870378.1 | |||
| AK002137.1 | BAA92101.1 | |||
| AK310397.1 | ||||
| BC005202.1 | AAH05202.1 | |||
| BC017914.1 | AAH17914.1 | |||
| BX647652.1 | ||||
| DA346124.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891706 | Chr.9:104480016 - 105304573 on Build GRCh38 | Gain |
|
| esv3621343 | Chr.9:104761673 - 104776613 on Build GRCh38 | Loss |
|
| nsv521783 | Chr.9:104757168 - 104770036 on Build GRCh38 | Loss |
|
| esv3891705 | Chr.9:104391828 - 106660921 on Build GRCh38 | Loss |
|

Genomic Map