Assay Details
Target Gene Details
Entrez Gene ID: | 6631 |
Gene Name: | small nuclear ribonucleoprotein polypeptide C |
Gene Aliases: |
U1C, Yhc1 |
Location: |
Chr.6:34757094-34773857 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNRPC | NM_003093.2 | NP_003084.1 | ||
| NR_029472.1 | ||||
| AI356808.1 | ||||
| AK223098.1 | BAD96818.1 | |||
| AK311955.1 | ||||
| BC121082.1 | ||||
| BC121083.1 | ||||
| BU153661.1 | ||||
| BU508721.1 | ||||
| CD109745.1 | ||||
| M18465.1 | AAA36618.1 | |||
| X12517.1 | CAA31037.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462905 | Chr.6:34760476 - 34918659 on Build GRCh38 | Gain |
|
| dgv5955n100 | Chr.6:34755946 - 34919539 on Build GRCh38 | Gain |
|
| esv2763967 | Chr.6:34598478 - 34957686 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map