Assay Details
Target Gene Details
Entrez Gene ID: | 222234 |
Gene Name: | family with sequence similarity 185 member A |
Gene Aliases: |
- |
Location: |
Chr.7:102748944-102913394 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM185A | XM_006715896.3 | XP_006715959.1 | ||
| XM_011515922.2 | XP_011514224.1 | |||
| XM_011515923.2 | XP_011514225.1 | |||
| XM_011515924.2 | XP_011514226.1 | |||
| XM_011515925.2 | XP_011514227.1 | |||
| XM_011515926.2 | XP_011514228.1 | |||
| XM_017011846.1 | XP_016867335.1 | |||
| XM_017011847.1 | XP_016867336.1 | |||
| XM_017011848.1 | XP_016867337.1 | |||
| XM_017011849.1 | XP_016867338.1 | |||
| AK297455.1 |
Target Gene Details
Entrez Gene ID: | 222235 |
Gene Name: | F-box and leucine rich repeat protein 13 |
Gene Aliases: |
DRC6, Fbl13 |
Location: |
Chr.7:102699226-103074841 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXL13 | NM_001111038.1 | 19 | 2376 | NP_001104508.1 |
| NM_001287150.1 | 19 | 2427 | NP_001274079.1 | |
| NM_145032.3 | 20 | 2688 | NP_659469.3 | |
| NR_105043.1 | 21 | 2525 | ||
| XM_005250205.3 | 21 | 2594 | XP_005250262.1 | |
| XM_005250207.3 | 20 | 2495 | XP_005250264.1 | |
| XM_005250208.3 | XP_005250265.1 | |||
| XM_005250209.2 | XP_005250266.1 | |||
| XM_006715898.2 | XP_006715961.1 | |||
| XM_011515930.2 | XP_011514232.1 | |||
| XM_017011850.1 | XP_016867339.1 | |||
| XM_017011852.1 | XP_016867341.1 | |||
| XM_017011853.1 | XP_016867342.1 | |||
| AK095387.1 | 15 | 1911 | BAC04540.1 | |
| AK097537.1 | BAC05092.1 | |||
| AL713709.1 | CAD28506.1 | |||
| AY359238.1 | AAR13262.1 | |||
| AY359239.1 | AAR13263.1 | |||
| BC020572.2 | AAH20572.2 | |||
| BC020575.2 | AAH20575.2 | |||
| BC026121.1 | AAH26121.1 | |||
| BC031285.1 | AAH31285.1 | |||
| BP378950.1 | 1 | 250 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761100 | Chr.7:102695340 - 102841531 on Build GRCh38 | Gain+Loss |
|
| nsv469620 | Chr.7:102655945 - 102813616 on Build GRCh38 | Loss |
|
| nsv608057 | Chr.7:102799737 - 102898068 on Build GRCh38 | Loss |
|
| esv3614358 | Chr.7:102794927 - 102853299 on Build GRCh38 | Gain |
|
| nsv470382 | Chr.7:102735216 - 102898068 on Build GRCh38 | Loss |
|
| dgv6538n100 | Chr.7:102171110 - 103063688 on Build GRCh38 | Gain |
|
| esv2752138 | Chr.7:102717873 - 103146688 on Build GRCh38 | Loss |
|

Genomic Map