Assay Details
Target Gene Details
Entrez Gene ID: | 51808 |
Gene Name: | phosphorylated adaptor for RNA export |
Gene Aliases: |
RNUXA |
Location: |
Chr.5:126600915-126627252 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHAX | NM_032177.3 | 5 | 1567 | NP_115553.2 |
| AK023255.1 | 5 | 1567 | BAB14489.1 | |
| AK024065.1 | 5 | 1535 | BAB14809.1 | |
| N44678.1 | 1 | 129 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462442 | Chr.5:126586076 - 126753498 on Build GRCh38 | Gain |
|
| nsv1030016 | Chr.5:126553411 - 126636590 on Build GRCh38 | Gain |
|
| esv3606679 | Chr.5:126424914 - 126763280 on Build GRCh38 | Gain |
|
| esv3606680 | Chr.5:126475605 - 126628219 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map