Assay Details
Target Gene Details
Entrez Gene ID: | 10318 |
Gene Name: | TNFAIP3 interacting protein 1 |
Gene Aliases: |
ABIN-1, NAF1, VAN, nip40-1 |
Location: |
Chr.5:151029943-151087660 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526224 | Chr.5:151003841 - 151070675 on Build GRCh38 | Gain |
|
| nsv600004 | Chr.5:151048634 - 151059427 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map