Assay Details
Target Gene Details
Entrez Gene ID: | 64744 |
Gene Name: | small ArfGAP2 |
Gene Aliases: |
SMAP1L |
Location: |
Chr.1:40373706-40423326 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMAP2 | NM_022733.2 | 1 | 437 | NP_073570.1 |
| XM_011541960.1 | 1 | 381 | XP_011540262.1 | |
| XM_017002093.1 | 1 | 435 | XP_016857582.1 | |
| AK313105.1 | 1 | 384 | ||
| AL137764.1 | CAB70912.1 | |||
| BC021133.1 | 1 | 87 | AAH21133.1 | |
| DA082592.1 |
More Information
Set Membership: |
|

Genomic Map