Assay Details
Target Gene Details
Entrez Gene ID: | 166378 |
Gene Name: | spermatogenesis associated 5 |
Gene Aliases: |
AFG2, EHLMRS, SPAF |
Location: |
Chr.4:122923070-123319450 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPATA5 | NM_001317799.1 | NP_001304728.1 | ||
| NM_145207.2 | NP_660208.2 | |||
| XM_005262783.4 | XP_005262840.1 | |||
| XM_011531678.2 | XP_011529980.1 | |||
| XM_011531679.2 | XP_011529981.1 | |||
| XM_017007825.1 | XP_016863314.1 | |||
| XM_017007826.1 | XP_016863315.1 | |||
| XM_017007827.1 | XP_016863316.1 | |||
| XM_017007828.1 | XP_016863317.1 | |||
| XM_017007830.1 | XP_016863319.1 | |||
| AF361489.1 | AAM00262.1 | |||
| AF479656.1 | AAM43608.1 | |||
| AK091384.1 | BAC03651.1 | |||
| AL832175.1 | ||||
| BC048217.1 | AAH48217.1 | |||
| DA619323.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3894022 | Chr.4:122865924 - 123123191 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map