Assay Details
Target Gene Details
Entrez Gene ID: | 2138 |
Gene Name: | EYA transcriptional coactivator and phosphatase 1 |
Gene Aliases: |
BOP, BOR, BOS1, OFC1 |
Location: |
Chr.8:71197433-71548130 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EYA1 | NM_000503.5 | 18 | 2557 | NP_000494.2 |
NM_001288574.1 | 18 | 2539 | NP_001275503.1 | |
NM_001288575.1 | 18 | 2475 | NP_001275504.1 | |
NM_172058.3 | 17 | 2149 | NP_742055.1 | |
NM_172059.3 | 16 | 1866 | NP_742056.1 | |
NM_172060.3 | 16 | 2021 | NP_742057.1 | |
XM_011517483.2 | 17 | 6854 | XP_011515785.1 | |
XM_011517484.2 | 19 | 2300 | XP_011515786.2 | |
XM_017013201.1 | 19 | 2343 | XP_016868690.1 | |
XM_017013202.1 | 19 | 2187 | XP_016868691.1 | |
XM_017013203.1 | 19 | 2312 | XP_016868692.1 | |
XM_017013204.1 | 19 | 2298 | XP_016868693.1 | |
XM_017013206.1 | 20 | 2296 | XP_016868695.1 | |
XM_017013207.1 | 18 | 2224 | XP_016868696.1 | |
XM_017013208.1 | 18 | 2579 | XP_016868697.1 | |
XM_017013209.1 | 18 | 2099 | XP_016868698.1 | |
XM_017013210.1 | 18 | 2208 | XP_016868699.1 | |
XM_017013211.1 | 18 | 2187 | XP_016868700.1 | |
XM_017013212.1 | 17 | 2454 | XP_016868701.1 | |
XM_017013213.1 | 17 | 6992 | XP_016868702.1 | |
AJ000097.1 | 17 | 2094 | CAA03922.1 | |
AJ000098.1 | 18 | 2555 | CAA03923.1 | |
AK127798.1 | 18 | 1975 | ||
AK295897.1 | 18 | 2048 | ||
BC121798.1 | ||||
BC121799.1 | ||||
BU679404.1 | 1 | 249 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
