Assay Details
Target Gene Details
| Entrez Gene ID: | 10780 | 
| Gene Name: | zinc finger protein 234 | 
| Gene Aliases: | HZF4, ZNF269 | 
| Location: | Chr.19:44141530-44160309 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 5 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| ZNF234 | NM_001144824.1 | 6 | 2210 | NP_001138296.1 | 
| NM_006630.2 | 6 | 2328 | NP_006621.1 | |
| XM_006722974.3 | 6 | 2469 | XP_006723037.1 | |
| XM_017026149.1 | 6 | 2439 | XP_016881638.1 | |
| XM_017026150.1 | 6 | 2227 | XP_016881639.1 | |
| XM_017026151.1 | 5 | 2202 | XP_016881640.1 | |
| XM_017026152.1 | 7 | 2071 | XP_016881641.1 | |
| XM_017026153.1 | 6 | 2683 | XP_016881642.1 | |
| AK289843.1 | 6 | 2270 | ||
| AK314064.1 | 6 | 2210 | ||
| BC007303.1 | 6 | 2260 | AAH07303.1 | |
| X78927.1 | 3 | 2007 | CAA55527.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv833840 | Chr.19:44016468 - 44211607 on Build GRCh38 | Gain |  ZNF224  ZNF226  ZNF223  ZNF234  ZNF222  ZNF227  ZNF225  LOC100379224  ZNF284 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top