Assay Details
Target Gene Details
Entrez Gene ID: | 661 |
Gene Name: | RNA polymerase III subunit D |
Gene Aliases: |
BN51T, RPC4, RPC53, TSBN51 |
Location: |
Chr.8:22245104-22251167 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR3D | NM_001722.2 | NP_001713.2 | ||
| AK026588.1 | ||||
| M17754.1 | AAA51838.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470205 | Chr.8:22191277 - 22256052 on Build GRCh38 | Loss |
|
| dgv12054n54 | Chr.8:22045805 - 22251098 on Build GRCh38 | Loss |
|
| nsv831265 | Chr.8:22199688 - 22366918 on Build GRCh38 | Loss |
|
| nsv831264 | Chr.8:22093700 - 22328040 on Build GRCh38 | Gain+Loss |
|
| esv2759604 | Chr.8:20675533 - 23254096 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79812935] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map