Assay Details
Target Gene Details
Entrez Gene ID: | 90580 |
Gene Name: | chromosome 19 open reading frame 52 |
Gene Aliases: |
- |
Location: |
Chr.19:10928748-10930241 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C19orf52 | NM_138358.2 | 2 | 1039 | NP_612367.1 |
| BC005075.1 | 2 | 1016 | AAH05075.2 | |
| BC007244.1 | 2 | 1029 | AAH07244.2 | |
| BC011833.2 | 2 | 1039 | AAH11833.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833749 | Chr.19:10881255 - 11027578 on Build GRCh38 | Loss |
|

Genomic Map