Assay Details
Target Gene Details
Entrez Gene ID: | 4798 |
Gene Name: | nuclear factor related to kappaB binding protein |
Gene Aliases: |
INO80G |
Location: |
Chr.11:129863773-129895596 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFRKB | NM_001143835.1 | 27 | 4296 | NP_001137307.1 |
| NM_006165.3 | 25 | 4291 | NP_006156.2 | |
| XM_005271567.3 | 27 | 4332 | XP_005271624.1 | |
| XM_005271568.3 | 26 | 4247 | XP_005271625.1 | |
| XM_011542850.2 | 28 | 4369 | XP_011541152.1 | |
| XM_011542851.2 | 27 | 4351 | XP_011541153.1 | |
| XM_011542852.2 | 26 | 4189 | XP_011541154.1 | |
| XM_017017793.1 | 26 | 4243 | XP_016873282.1 | |
| XM_017017794.1 | 26 | 4207 | XP_016873283.1 | |
| XM_017017795.1 | 28 | 4333 | XP_016873284.1 | |
| XM_017017796.1 | 26 | 4211 | XP_016873285.1 | |
| XM_017017797.1 | 27 | 3976 | XP_016873286.1 | |
| XM_017017798.1 | 28 | 4029 | XP_016873287.1 | |
| XM_017017799.1 | 28 | 4443 | XP_016873288.1 | |
| XM_017017800.1 | 28 | 4383 | XP_016873289.1 | |
| XM_017017801.1 | 13 | 3343 | XP_016873290.1 | |
| AL512730.1 | 9 | 2134 | CAC21663.1 | |
| BC063280.1 | 27 | 4242 | AAH63280.1 | |
| X80878.1 | 25 | 4300 | CAA56846.1 |
More Information
Set Membership: |
|

Genomic Map