Assay Details
Target Gene Details
Entrez Gene ID: | 403282 |
Gene Name: | olfactory receptor family 6 subfamily C member 65 |
Gene Aliases: |
- |
Location: |
Chr.12:55400529-55401467 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6C65 | NM_001005518.1 | 1 | 507 | NP_001005518.1 |
| BC136897.1 | 1 | 606 | ||
| BC136899.1 | 1 | 606 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045950 | Chr.12:55360325 - 55440504 on Build GRCh38 | Gain |
|
| nsv1046651 | Chr.12:55385158 - 55430198 on Build GRCh38 | Gain |
|
| nsv559001 | Chr.12:55113681 - 55663924 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map