Assay Details
Target Gene Details
Entrez Gene ID: | 146059 |
Gene Name: | codanin 1 |
Gene Aliases: |
CDA1, CDAI, CDAN1A, DLT, PRO1295 |
Location: |
Chr.15:42723557-42737219 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDAN1 | NM_138477.2 | 28 | 3871 | NP_612486.2 |
| XM_005254176.4 | 28 | 3786 | XP_005254233.1 | |
| XM_011521270.2 | 28 | 3811 | XP_011519572.1 | |
| XM_011521271.2 | 28 | 3808 | XP_011519573.1 | |
| XM_011521274.2 | 25 | 2857 | XP_011519576.1 | |
| AF525398.1 | 28 | 3868 | AAO14994.1 | |
| AL133019.1 | 1 | 1217 | ||
| BC001092.2 | 20 | 2326 | AAH01092.1 | |
| BC008333.1 | 20 | 2326 | AAH08333.1 | |
| BC008334.1 | 20 | 2326 | AAH08334.1 | |
| BC052568.1 | 28 | 3759 | AAH52568.1 | |
| BC066640.1 | 28 | 3759 | AAH66640.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528725 | Chr.15:42463357 - 42812208 on Build GRCh38 | Gain |
|
| nsv952615 | Chr.15:42714903 - 42736802 on Build GRCh38 | Deletion |
|
| nsv457122 | Chr.15:42656664 - 42770350 on Build GRCh38 | Loss |
|
| nsv517553 | Chr.15:42678701 - 42731284 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map