Assay Details
Target Gene Details
Entrez Gene ID: | 284273 |
Gene Name: | zinc binding alcohol dehydrogenase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.18:75195108-75209348 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZADH2 | NM_001306093.1 | 2 | 904 | NP_001293022.1 |
| NM_175907.5 | 2 | 1082 | NP_787103.1 | |
| AK292880.1 | 2 | 837 | ||
| AK302801.1 | 2 | 634 | ||
| BC018081.1 | 2 | 1060 | AAH18081.1 | |
| BC033780.1 | 2 | 968 | AAH33780.1 | |
| BC078661.1 | 2 | 895 | AAH78661.1 | |
| BX647730.1 | 2 | 905 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3643133 | Chr.18:75067524 - 75744229 on Build GRCh38 | Loss |
|
| nsv833695 | Chr.18:75082974 - 75277673 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map