Assay Details
Target Gene Details
Entrez Gene ID: | 6518 |
Gene Name: | solute carrier family 2 member 5 |
Gene Aliases: |
GLUT-5, GLUT5 |
Location: |
Chr.1:9036941-9093407 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC2A5 | NM_001135585.1 | 4 | 621 | NP_001129057.1 |
| NM_003039.2 | 4 | 621 | NP_003030.1 | |
| XM_005263491.3 | 5 | 419 | XP_005263548.1 | |
| XM_017002133.1 | 6 | 647 | XP_016857622.1 | |
| XM_017002134.1 | 6 | 613 | XP_016857623.1 | |
| XM_017002135.1 | 6 | 707 | XP_016857624.1 | |
| XM_017002136.1 | 6 | 683 | XP_016857625.1 | |
| XM_017002137.1 | 7 | 719 | XP_016857626.1 | |
| XM_017002138.1 | 7 | 695 | XP_016857627.1 | |
| XM_017002139.1 | 5 | 629 | XP_016857628.1 | |
| XM_017002140.1 | 6 | 550 | XP_016857629.1 | |
| XM_017002141.1 | 5 | 452 | XP_016857630.1 | |
| XM_017002142.1 | 2 | 192 | XP_016857631.1 | |
| AF451839.1 | 2 | 134 | ||
| AF479408.1 | 2 | 134 | AAL87107.1 | |
| AK289849.1 | 4 | 377 | ||
| AK290398.1 | 4 | 411 | ||
| AK294368.1 | 4 | 407 | ||
| AK295785.1 | ||||
| AK300091.1 | 2 | 78 | ||
| AK300477.1 | 2 | 197 | ||
| AK302047.1 | 4 | 262 | ||
| AK302057.1 | 7 | 545 | ||
| AK302137.1 | ||||
| BC001692.2 | 4 | 380 | AAH01692.1 | |
| BC001820.2 | 4 | 380 | AAH01820.1 | |
| BC035878.1 | 4 | 408 | AAH35878.1 | |
| M55531.1 | 4 | 404 | AAA52570.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585161 | Chr.1:9045371 - 9049438 on Build GRCh38 | Gain |
|
| nsv1004025 | Chr.1:9000983 - 9069086 on Build GRCh38 | Gain |
|
| esv3577721 | Chr.1:9044393 - 9059929 on Build GRCh38 | Loss |
|
| nsv545347 | Chr.1:9020352 - 9072997 on Build GRCh38 | Loss |
|
| nsv831981 | Chr.1:9031658 - 9187508 on Build GRCh38 | Loss |
|
| dgv15n100 | Chr.1:8992095 - 9056684 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map