Assay Details
Target Gene Details
Entrez Gene ID: | 2585 |
Gene Name: | galactokinase 2 |
Gene Aliases: |
GK2 |
Location: |
Chr.15:49155757-49367883 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALK2 | NM_001001556.2 | NP_001001556.1 | ||
| NM_001289030.1 | 1 | 264 | NP_001275959.1 | |
| NM_001289031.1 | 1 | 264 | NP_001275960.1 | |
| NM_002044.3 | 1 | 264 | NP_002035.1 | |
| XM_005254279.3 | 1 | 53 | XP_005254336.1 | |
| XM_005254280.3 | XP_005254337.1 | |||
| XM_005254284.3 | XP_005254341.1 | |||
| XM_006720461.3 | 1 | 53 | XP_006720524.1 | |
| XM_006720463.3 | XP_006720526.1 | |||
| XM_006720464.3 | XP_006720527.1 | |||
| XM_011521441.2 | 1 | 129 | XP_011519743.1 | |
| XM_017022063.1 | XP_016877552.1 | |||
| XM_017022065.1 | XP_016877554.1 | |||
| XM_017022066.1 | 1 | 202 | XP_016877555.1 | |
| AF461816.1 | AAP97708.1 | |||
| AK098049.1 | ||||
| AK301361.1 | ||||
| AK316440.1 | ||||
| AL832745.1 | ||||
| BC005141.2 | AAH05141.1 | |||
| BI766538.1 | 1 | 55 | ||
| BQ921903.1 | ||||
| DB457948.1 | 1 | 264 | ||
| DC298656.1 | 1 | 253 | ||
| DC390737.1 | ||||
| M84443.1 | AAA58612.1 |
More Information
Set Membership: |
|

Genomic Map