Assay Details
Target Gene Details
Entrez Gene ID: | 5886 |
Gene Name: | RAD23 homolog A, nucleotide excision repair protein |
Gene Aliases: |
HHR23A, HR23A |
Location: |
Chr.19:12945814-12953643 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD23A | NM_001270362.1 | NP_001257291.1 | ||
| NM_001270363.1 | NP_001257292.1 | |||
| NM_005053.3 | NP_005044.1 | |||
| NR_072976.1 | ||||
| AB209713.1 | BAD92950.1 | |||
| AK289908.1 | ||||
| AK293219.1 | ||||
| BC014026.2 | AAH14026.1 | |||
| BC088364.1 | AAH88364.1 | |||
| BX448989.2 | ||||
| D21235.1 | BAA04767.1 | |||
| M77024.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion |
|
| nsv833754 | Chr.19:12876208 - 13028186 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map