Assay Details
Target Gene Details
Entrez Gene ID: | 116071 |
Gene Name: | basic leucine zipper ATF-like transcription factor 2 |
Gene Aliases: |
SARI |
Location: |
Chr.11:64987945-64997045 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BATF2 | NM_001300807.1 | 2 | 837 | NP_001287736.1 |
| NM_001300808.1 | 2 | 718 | NP_001287737.1 | |
| NM_138456.3 | 3 | 905 | NP_612465.3 | |
| AK092453.1 | 2 | 715 | ||
| AK096702.1 | 3 | 905 | BAC04846.1 | |
| AK300917.1 | 2 | 834 | ||
| BC012330.1 | 2 | 695 | AAH12330.1 | |
| HM134154.1 | 3 | 905 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555208 | Chr.11:64877647 - 65003707 on Build GRCh38 | Loss |
|
| nsv526181 | Chr.11:64870569 - 65489858 on Build GRCh38 | Gain |
|
| esv32668 | Chr.11:64975598 - 64990615 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map