Assay Details
Target Gene Details
Entrez Gene ID: | 56936 |
Gene Name: | coiled-coil domain containing 177 |
Gene Aliases: |
C14orf162, PLPL |
Location: |
Chr.14:69569814-69574883 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC177 | NM_001271507.1 | 2 | 2728 | NP_001258436.1 |
| AK312175.1 | 1 | 51 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564970 | Chr.14:69533682 - 69573444 on Build GRCh38 | Loss |
|
| nsv1335 | Chr.14:69569409 - 69595867 on Build GRCh38 | Deletion |
|
| dgv3743n54 | Chr.14:69568783 - 69574646 on Build GRCh38 | Loss |
|

Genomic Map