Assay Details
Target Gene Details
Entrez Gene ID: | 5611 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C3 |
Gene Aliases: |
ACPHD, ERdj6, HP58, P58, P58IPK, PRKRI |
Location: |
Chr.13:95677139-95794989 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC3 | NM_006260.4 | NP_006251.1 | ||
| XM_011521104.2 | XP_011519406.1 | |||
| XM_011521105.1 | XP_011519407.1 | |||
| XM_017020674.1 | XP_016876163.1 | |||
| AK292947.1 | ||||
| AK312861.1 | ||||
| BC047936.1 | AAH47936.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832690 | Chr.13:95678503 - 95855823 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map