Assay Details
Target Gene Details
Entrez Gene ID: | 116443 |
Gene Name: | glutamate ionotropic receptor NMDA type subunit 3A |
Gene Aliases: |
GluN3A, NMDAR-L, NR3A |
Location: |
Chr.9:101569352-101738580 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIN3A | NM_133445.2 | NP_597702.2 | ||
| AB075853.1 | ||||
| AF416558.1 | AAL40734.1 | |||
| AJ416950.2 | CAC95229.2 | |||
| AK127342.1 | ||||
| BC132866.1 | ||||
| BC132868.1 | ||||
| BC167432.1 |
Target Gene Details
Entrez Gene ID: | 5535 |
Gene Name: | protein phosphatase 3 regulatory subunit B, beta |
Gene Aliases: |
PPP3RL |
Location: |
Chr.9:101591615-101595001 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP3R2 | NM_147180.3 | 1 | 364 | NP_671709.1 |
| AF085237.1 | 1 | 284 | AAL40395.1 | |
| AF145026.1 | 1 | 284 | AAP97278.1 | |
| AF400667.1 | 1 | 338 | AAP57772.1 | |
| AK057524.1 | 1 | 323 | BAB71521.1 | |
| BC030595.2 | 1 | 364 | AAH30595.1 | |
| BC066299.1 | 1 | 305 | AAH66299.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831672 | Chr.9:101538473 - 101718567 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map