Assay Details
Target Gene Details
Entrez Gene ID: | 54757 |
Gene Name: | family with sequence similarity 20 member A |
Gene Aliases: |
AI1G, AIGFS, FP2747 |
Location: |
Chr.17:68534561-68600954 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM20A | NM_001243746.1 | NP_001230675.1 | ||
| NM_017565.3 | NP_060035.2 | |||
| NR_027751.1 | ||||
| XM_006721959.2 | XP_006722022.1 | |||
| XM_011524918.2 | XP_011523220.1 | |||
| XM_017024781.1 | XP_016880270.1 | |||
| AB545606.1 | ||||
| AK056789.1 | BAB71285.1 | |||
| AK298071.1 | ||||
| AL133105.1 | CAB61412.1 | |||
| AY358197.1 | AAQ88564.1 | |||
| BC036222.1 | AAH36222.1 | |||
| BC136686.1 | ||||
| BC136689.1 | ||||
| DB057818.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833524 | Chr.17:68527889 - 68709053 on Build GRCh38 | Gain |
|
| nsv510723 | Chr.17:68533527 - 68561284 on Build GRCh38 | Deletion |
|
| nsv522126 | Chr.17:68542098 - 68552316 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map