| nsv482937 | 
						
							
								Chr.1:10001 - 2368561
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 PLEKHN1
							
								 UBE2J2
							
								 LOC105378589
							
								 CFAP74
							
								 SDF4
							
								 AGRN
							
								 CPTP
							
								 MTND2P28
							
								 LOC100287934
							
								 LOC105378949
							
								 RNF223
							
								 ATAD3C
							
								 C1orf159
							
								 B3GALT6
							
								 SKI
							
								 TNFRSF18
							
								 LOC100506504
							
								 CPSF3L
							
								 MORN1
							
								 CDK11A
							
								 MIR6727
							
								 CDK11B
							
								 LOC100288175
							
								 MIB2
							
								 LOC107985728
							
								 MMP23B
							
								 LOC284600
							
								 SAMD11
							
								 GNB1
							
								 TMEM52
							
								 SSU72
							
								 LINC01128
							
								 MIR200A
							
								 DDX11L1
							
								 LOC101928626
							
								 MRPL20
							
								 MIR6726
							
								 MTND1P23
							
								 OR4F16
							
								 PERM1
							
								 ACAP3
							
								 NOC2L
							
								 TTLL10
							
								 LOC100288069
							
								 MIR6859-2
							
								 TTLL10-AS1
							
								 SLC35E2
							
								 HES4
							
								 LOC105378591
							
								 MIR200B
							
								 FAM41C
							
								 CCNL2
							
								 LOC100129534
							
								 ISG15
							
								 LOC102724312
							
								 MIR6808
							
								 FAM138A
							
								 FNDC10
							
								 TNFRSF4
							
								 AURKAIP1
							
								 DVL1
							
								 LOC100134822
							
								 TAS1R3
							
								 OR4F29
							
								 LOC148413
							
								 SLC35E2B
							
								 LOC105378592
							
								 LOC102725121
							
								 LINC00115
							
								 GABRD
							
								 TMEM240
							
								 MMP23A
							
								 OR4F5
							
								 MIR6723
							
								 SCNN1D
							
								 ATAD3A
							
								 LOC105378948
							
								 LINC01342
							
								 ATAD3B
							
								 WASH7P
							
								 KLHL17
							
								 FAAP20
							
								 LOC107985729
							
								 TMEM88B
							
								 VWA1
							
								 NADK
							
								 MXRA8
							
								 CALML6
							
								 ANKRD65
							
								 FAM87B
							
								 MIR1302-2
							
								 LOC105378947
							
								 LOC729737
							
								 PRKCZ
							
								 LOC100130417
							
								 LOC107984841
							
								 PUSL1
							
								 FAM132A
							
								 MIR6859-1
							
								 LOC100132287
							
								 MIR429
							
								 LOC100133331
							
						 | 
					
				
		    		
						| nsv1003578 | 
						
							
								Chr.1:1068249 - 1282706
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 MIR200B
							
								 MIR200A
							
								 C1orf159
							
								 LINC01342
							
								 UBE2J2
							
								 B3GALT6
							
								 TNFRSF18
							
								 SDF4
							
								 FAM132A
							
								 SCNN1D
							
								 RNF223
							
								 TNFRSF4
							
								 TTLL10
							
								 LOC105378948
							
								 TTLL10-AS1
							
								 MIR429
							
						 | 
					
				
		    		
						| nsv470680 | 
						
							
								Chr.1:1070426 - 1378635
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 MIR200B
							
								 LINC01342
							
								 UBE2J2
							
								 SDF4
							
								 CPTP
							
								 MIR6808
							
								 MXRA8
							
								 RNF223
							
								 TNFRSF4
							
								 AURKAIP1
							
								 DVL1
							
								 MIR200A
							
								 C1orf159
							
								 TAS1R3
							
								 B3GALT6
							
								 TNFRSF18
							
								 PUSL1
							
								 MIR6726
							
								 FAM132A
							
								 CPSF3L
							
								 ACAP3
							
								 SCNN1D
							
								 MIR6727
							
								 TTLL10
							
								 TTLL10-AS1
							
								 MIR429
							
						 | 
					
				
		    		
						| nsv544956 | 
						
							
								Chr.1:1171093 - 1241217
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 B3GALT6
							
								 TNFRSF4
							
								 TTLL10
							
								 TNFRSF18
							
								 SDF4
							
								 TTLL10-AS1
							
						 | 
					
				
		    		
						| nsv544963 | 
						
							
								Chr.1:1206945 - 1214281
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 TNFRSF4
							
								 TNFRSF18
							
						 | 
					
				
		    		
						| nsv1160772 | 
						
							
								Chr.1:1027585 - 1333467
							    on Build GRCh38
						
							
						 | 
						Duplication | 
						
							
								 MIR200B
							
								 LINC01342
							
								 UBE2J2
							
								 LOC100288175
							
								 SDF4
							
								 AGRN
							
								 CPTP
							
								 RNF223
							
								 TNFRSF4
							
								 MIR200A
							
								 C1orf159
							
								 TAS1R3
							
								 B3GALT6
							
								 TNFRSF18
							
								 PUSL1
							
								 MIR6726
							
								 FAM132A
							
								 CPSF3L
							
								 ACAP3
							
								 SCNN1D
							
								 MIR6727
							
								 TTLL10
							
								 LOC105378948
							
								 TTLL10-AS1
							
								 MIR429
							
						 | 
					
				
		    		
						| nsv428334 | 
						
							
								Chr.1:874371 - 1220569
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 PLEKHN1
							
								 MIR200B
							
								 FAM41C
							
								 LINC01342
							
								 LOC100288175
							
								 KLHL17
							
								 SDF4
							
								 LOC107985728
							
								 AGRN
							
								 ISG15
							
								 LOC284600
							
								 SAMD11
							
								 RNF223
							
								 TNFRSF4
							
								 MIR200A
							
								 C1orf159
							
								 LOC100130417
							
								 TNFRSF18
							
								 PERM1
							
								 NOC2L
							
								 TTLL10
							
								 LOC105378948
							
								 TTLL10-AS1
							
								 MIR429
							
								 HES4
							
						 | 
					
				
		    		
						| nsv10161 | 
						
							
								Chr.1:776731 - 1777210
							    on Build GRCh38
						
							
						 | 
						Gain+Loss | 
						
							
								 PLEKHN1
							
								 MIR200B
							
								 FAM41C
							
								 UBE2J2
							
								 SDF4
							
								 CCNL2
							
								 AGRN
							
								 ISG15
							
								 CPTP
							
								 LOC102724312
							
								 MIR6808
							
								 LOC100287934
							
								 FNDC10
							
								 RNF223
							
								 TNFRSF4
							
								 AURKAIP1
							
								 DVL1
							
								 ATAD3C
							
								 C1orf159
							
								 TAS1R3
							
								 LOC148413
							
								 SLC35E2B
							
								 B3GALT6
							
								 LINC00115
							
								 TNFRSF18
							
								 TMEM240
							
								 CPSF3L
							
								 MMP23A
							
								 CDK11A
							
								 SCNN1D
							
								 MIR6727
							
								 ATAD3A
							
								 CDK11B
							
								 LOC105378948
							
								 LINC01342
							
								 ATAD3B
							
								 LOC100288175
							
								 KLHL17
							
								 MIB2
							
								 LOC107985729
							
								 LOC107985728
							
								 TMEM88B
							
								 MMP23B
							
								 LOC284600
							
								 VWA1
							
								 SAMD11
							
								 NADK
							
								 MXRA8
							
								 ANKRD65
							
								 FAM87B
							
								 SSU72
							
								 LINC01128
							
								 MIR200A
							
								 LOC100130417
							
								 PUSL1
							
								 MRPL20
							
								 MIR6726
							
								 FAM132A
							
								 PERM1
							
								 ACAP3
							
								 NOC2L
							
								 TTLL10
							
								 LOC100288069
							
								 TTLL10-AS1
							
								 SLC35E2
							
								 MIR429
							
								 HES4
							
						 | 
					
				
		    		
						| nsv950452 | 
						
							
								Chr.1:1106921 - 1637538
							    on Build GRCh38
						
							
						 | 
						Deletion | 
						
							
								 MIR200B
							
								 LINC01342
							
								 UBE2J2
							
								 ATAD3B
							
								 MIB2
							
								 SDF4
							
								 LOC107985729
							
								 TMEM88B
							
								 CCNL2
							
								 MMP23B
							
								 CPTP
							
								 VWA1
							
								 LOC102724312
							
								 MIR6808
							
								 MXRA8
							
								 ANKRD65
							
								 FNDC10
							
								 TNFRSF4
							
								 AURKAIP1
							
								 DVL1
							
								 ATAD3C
							
								 SSU72
							
								 MIR200A
							
								 C1orf159
							
								 TAS1R3
							
								 LOC148413
							
								 B3GALT6
							
								 TNFRSF18
							
								 PUSL1
							
								 MRPL20
							
								 MIR6726
							
								 FAM132A
							
								 TMEM240
							
								 CPSF3L
							
								 ACAP3
							
								 SCNN1D
							
								 MIR6727
							
								 ATAD3A
							
								 TTLL10
							
								 CDK11B
							
								 TTLL10-AS1
							
								 MIR429
							
						 | 
					
				
		    		
						| nsv517709 | 
						
							
								Chr.1:817186 - 1275912
							    on Build GRCh38
						
							
						 | 
						Gain+Loss | 
						
							
								 PLEKHN1
							
								 MIR200B
							
								 FAM41C
							
								 LINC01342
							
								 UBE2J2
							
								 LOC100288175
							
								 KLHL17
							
								 SDF4
							
								 LOC107985728
							
								 AGRN
							
								 ISG15
							
								 LOC284600
							
								 SAMD11
							
								 FAM87B
							
								 RNF223
							
								 TNFRSF4
							
								 LINC01128
							
								 MIR200A
							
								 C1orf159
							
								 LOC100130417
							
								 B3GALT6
							
								 LINC00115
							
								 TNFRSF18
							
								 FAM132A
							
								 PERM1
							
								 NOC2L
							
								 TTLL10
							
								 LOC105378948
							
								 TTLL10-AS1
							
								 MIR429
							
								 HES4
							
						 | 
					
				
		    		
						| nsv1160644 | 
						
							
								Chr.1:939652 - 1220663
							    on Build GRCh38
						
							
						 | 
						Deletion | 
						
							
								 PLEKHN1
							
								 MIR200B
							
								 MIR200A
							
								 C1orf159
							
								 LINC01342
							
								 LOC100288175
							
								 KLHL17
							
								 TNFRSF18
							
								 SDF4
							
								 AGRN
							
								 ISG15
							
								 SAMD11
							
								 PERM1
							
								 NOC2L
							
								 RNF223
							
								 TNFRSF4
							
								 TTLL10
							
								 LOC105378948
							
								 TTLL10-AS1
							
								 MIR429
							
								 HES4
							
						 | 
					
				
		    		
						| dgv2n67 | 
						
							
								Chr.1:877618 - 1426500
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 PLEKHN1
							
								 MIR200B
							
								 LINC01342
							
								 UBE2J2
							
								 LOC100288175
							
								 KLHL17
							
								 SDF4
							
								 LOC107985728
							
								 TMEM88B
							
								 CCNL2
							
								 AGRN
							
								 ISG15
							
								 CPTP
							
								 LOC284600
							
								 SAMD11
							
								 MIR6808
							
								 MXRA8
							
								 ANKRD65
							
								 RNF223
							
								 TNFRSF4
							
								 AURKAIP1
							
								 DVL1
							
								 MIR200A
							
								 C1orf159
							
								 TAS1R3
							
								 LOC100130417
							
								 LOC148413
							
								 B3GALT6
							
								 TNFRSF18
							
								 PUSL1
							
								 MRPL20
							
								 MIR6726
							
								 FAM132A
							
								 CPSF3L
							
								 PERM1
							
								 ACAP3
							
								 NOC2L
							
								 SCNN1D
							
								 MIR6727
							
								 TTLL10
							
								 LOC105378948
							
								 TTLL10-AS1
							
								 MIR429
							
								 HES4
							
						 | 
					
				
		    		
						| dgv24n54 | 
						
							
								Chr.1:1205055 - 1213515
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 TNFRSF4
							
								 TNFRSF18
							
						 | 
					
				
		    		
						| nsv469848 | 
						
							
								Chr.1:1146082 - 1322235
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 MIR200B
							
								 MIR200A
							
								 UBE2J2
							
								 B3GALT6
							
								 TNFRSF18
							
								 SDF4
							
								 PUSL1
							
								 MIR6726
							
								 FAM132A
							
								 CPSF3L
							
								 ACAP3
							
								 SCNN1D
							
								 MIR6727
							
								 TNFRSF4
							
								 TTLL10
							
								 TTLL10-AS1
							
								 MIR429
							
						 |