Assay Details
Target Gene Details
Entrez Gene ID: | 6945 |
Gene Name: | MLX, MAX dimerization protein |
Gene Aliases: |
MAD7, MXD7, TCFL4, bHLHd13 |
Location: |
Chr.17:42567060-42573203 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLX | NM_170607.2 | 8 | 2104 | NP_733752.1 |
| NM_198204.1 | 8 | 1942 | NP_937847.1 | |
| NM_198205.1 | 7 | 1852 | NP_937848.1 | |
| XM_017024991.1 | XP_016880480.1 | |||
| AF203978.1 | AAF14638.1 | |||
| AF213666.1 | 7 | 1840 | AAG40145.1 | |
| AF213667.1 | 8 | 1930 | AAG40146.1 | |
| AF213668.1 | 8 | 2092 | AAG40147.1 | |
| AK000150.1 | BAA90977.1 | |||
| BC010689.2 | AAH10689.1 | |||
| BQ186038.1 | 1 | 455 |
Target Gene Details
Entrez Gene ID: | 29893 |
Gene Name: | PSMC3 interacting protein |
Gene Aliases: |
GT198, HOP2, HUMGT198A, ODG3, TBPIP |
Location: |
Chr.17:42572310-42577831 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMC3IP | NM_001256014.1 | 7 | 998 | NP_001242943.1 |
| NM_001256015.1 | 8 | 1155 | NP_001242944.1 | |
| NM_001256016.1 | 9 | 1041 | NP_001242945.1 | |
| NM_013290.6 | 8 | 986 | NP_037422.2 | |
| NM_016556.3 | 8 | 1022 | NP_057640.1 | |
| NR_045669.1 | 8 | 1302 | ||
| NR_045670.1 | 8 | 925 | ||
| NR_045671.1 | 7 | 901 | ||
| AB030304.1 | 8 | 920 | BAA92872.1 | |
| AK126369.1 | 6 | 1968 | ||
| BC008792.1 | 8 | 993 | AAH08792.1 | |
| U18921.1 | 2 | 341 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064012 | Chr.17:42427786 - 42606703 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map