Assay Details
Target Gene Details
Entrez Gene ID: | 9295 |
Gene Name: | serine and arginine rich splicing factor 11 |
Gene Aliases: |
NET2, SFRS11, dJ677H15.2, p54 |
Location: |
Chr.1:70205682-70252020 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950171 | Chr.1:70216518 - 70233517 on Build GRCh38 | Duplication |
|
| nsv1014292 | Chr.1:70223532 - 70333382 on Build GRCh38 | Gain |
|
| nsv1343 | Chr.1:70199486 - 70231317 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map