Assay Details
Target Gene Details
Entrez Gene ID: | 8672 |
Gene Name: | eukaryotic translation initiation factor 4 gamma 3 |
Gene Aliases: |
eIF-4G 3, eIF4G 3, eIF4GII |
Location: |
Chr.1:20806292-21177320 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 38 - Exon 38 |
More Information
Set Membership: |
|

Genomic Map