Assay Details
Target Gene Details
Entrez Gene ID: | 25906 |
Gene Name: | anaphase promoting complex subunit 15 |
Gene Aliases: |
APC15, C11orf51, HSPC020 |
Location: |
Chr.11:72106254-72112793 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANAPC15 | NM_001278485.1 | NP_001265414.1 | ||
| NM_001278486.1 | NP_001265415.1 | |||
| NM_001278487.1 | NP_001265416.1 | |||
| NM_001278488.1 | NP_001265417.1 | |||
| NM_001278489.1 | NP_001265418.1 | |||
| NM_001278490.1 | NP_001265419.1 | |||
| NM_014042.2 | NP_054761.1 | |||
| XM_017017488.1 | XP_016872977.1 | |||
| XM_017017491.1 | XP_016872980.1 | |||
| XM_017017493.1 | XP_016872982.1 | |||
| XM_017017494.1 | XP_016872983.1 | |||
| XM_017017495.1 | XP_016872984.1 | |||
| AF077206.1 | AAD27001.1 | |||
| AK222622.1 | ||||
| AK311146.1 | ||||
| AL080071.1 | CAB45698.1 | |||
| BC005156.2 | AAH05156.1 | |||
| BC005393.1 | AAH05393.1 | |||
| BG752082.1 | ||||
| BG764016.1 | ||||
| BU532056.1 | ||||
| BU930156.1 | ||||
| CN306434.1 |
More Information
Set Membership: |
|

Genomic Map