Assay Details
Target Gene Details
Entrez Gene ID: | 51655 |
Gene Name: | ras related dexamethasone induced 1 |
Gene Aliases: |
AGS1, DEXRAS1, MGC:26290 |
Location: |
Chr.17:17494437-17496395 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASD1 | NM_001199989.1 | 2 | 1572 | NP_001186918.1 |
| NM_016084.4 | 2 | 1646 | NP_057168.1 | |
| AF069506.1 | 2 | 1577 | AAD34206.1 | |
| AF177335.1 | 2 | 1625 | AAG17979.1 | |
| BC018041.1 | 2 | 1644 | AAH18041.1 |
More Information
Set Membership: |
|

Genomic Map