Assay Details
Target Gene Details
Entrez Gene ID: | 54539 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit B11 |
Gene Aliases: |
CI-ESSS, ESSS, LSDMCA3, NP17.3, Np15, P17.3 |
Location: |
Chr.X:47142216-47145210 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFB11 | NM_001135998.2 | 1 | 543 | NP_001129470.1 |
| NM_019056.6 | 1 | 543 | NP_061929.2 | |
| AF044213.1 | AAL32064.1 | |||
| AF134476.1 | AAP97253.1 | |||
| AF251063.1 | 1 | 374 | AAK34953.1 | |
| AK000501.1 | BAA91208.1 | |||
| AY359056.1 | 1 | 543 | AAQ89415.1 | |
| BC010665.1 | 1 | 351 | AAH10665.1 | |
| BC107805.1 | AAI07806.1 | |||
| CR457239.1 | CAG33520.1 |
More Information
Set Membership: |
|

Genomic Map