Assay Details
Target Gene Details
Entrez Gene ID: | 57452 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 16 |
Gene Aliases: |
GALNACT16, GALNTL1, GalNAc-T16 |
Location: |
Chr.14:69259546-69379904 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT16 | NM_001168368.1 | 1 | 360 | NP_001161840.1 |
| NM_020692.2 | 1 | 360 | NP_065743.2 | |
| XM_011537005.1 | 1 | 172 | XP_011535307.1 | |
| XM_011537006.2 | 1 | 172 | XP_011535308.1 | |
| XM_011537007.2 | 1 | 172 | XP_011535309.1 | |
| XM_017021498.1 | 2 | 347 | XP_016876987.1 | |
| AB032956.1 | 1 | 150 | BAA86444.1 | |
| AB078143.1 | 1 | 33 | BAD93178.1 | |
| AJ505951.1 | 1 | 33 | CAD44534.1 | |
| AK289745.1 | 1 | 119 | ||
| AK299588.1 | ||||
| AK315848.1 | 1 | 124 | ||
| BC036812.1 | 1 | 111 | ||
| BC098578.1 | 1 | 126 | AAH98578.1 | |
| BC122545.1 | 1 | 133 | ||
| DA493083.1 | 1 | 360 |
More Information
Set Membership: |
|

Genomic Map