Assay Details
Target Gene Details
Entrez Gene ID: | 4140 |
Gene Name: | microtubule affinity regulating kinase 3 |
Gene Aliases: |
CTAK1, KP78, PAR1A, Par-1a |
Location: |
Chr.14:103385364-103503831 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARK3 | NM_001128918.2 | 1 | 576 | NP_001122390.2 |
| NM_001128919.2 | 1 | 576 | NP_001122391.2 | |
| NM_001128920.2 | 1 | 576 | NP_001122392.2 | |
| NM_001128921.2 | 1 | 576 | NP_001122393.2 | |
| NM_002376.6 | 1 | 576 | NP_002367.5 | |
| XM_005267641.4 | XP_005267698.2 | |||
| XM_005267642.4 | 1 | 576 | XP_005267699.2 | |
| XM_005267643.4 | 1 | 576 | XP_005267700.2 | |
| XM_017021291.1 | 1 | 576 | XP_016876780.1 | |
| XM_017021292.1 | 1 | 576 | XP_016876781.1 | |
| XM_017021293.1 | XP_016876782.1 | |||
| XM_017021294.1 | 1 | 517 | XP_016876783.1 | |
| XM_017021295.1 | 1 | 532 | XP_016876784.1 | |
| XM_017021296.1 | 1 | 549 | XP_016876785.1 | |
| XM_017021297.1 | 1 | 525 | XP_016876786.1 | |
| XM_017021298.1 | 1 | 525 | XP_016876787.1 | |
| XM_017021299.1 | 1 | 576 | XP_016876788.1 | |
| XM_017021301.1 | 1 | 206 | XP_016876790.1 | |
| XM_017021306.1 | 1 | 576 | XP_016876795.1 | |
| XM_017021307.1 | 1 | 576 | XP_016876796.1 | |
| XM_017021308.1 | 1 | 576 | XP_016876797.1 | |
| AF465413.1 | 1 | 420 | AAL69982.1 | |
| AK307448.1 | ||||
| BC024773.1 | 1 | 399 | AAH24773.1 | |
| BX161395.1 | 1 | 506 | CAD61882.1 | |
| M80359.1 | 1 | 80 | AAA59991.1 | |
| U64205.1 | 1 | 284 | AAC15093.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9168 | Chr.14:103065392 - 103446969 on Build GRCh38 | Loss |
|
| esv3635592 | Chr.14:103373908 - 103388946 on Build GRCh38 | Gain |
|
| nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss |
|
| dgv422e214 | Chr.14:103355152 - 103437655 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map