Assay Details
Target Gene Details
Entrez Gene ID: | 27085 |
Gene Name: | MDM2 binding protein |
Gene Aliases: |
MDM2BP |
Location: |
Chr.8:120445398-120523635 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTBP | NM_022045.4 | 1 | 95 | NP_071328.2 |
| XM_011516962.2 | 1 | 92 | XP_011515264.1 | |
| XM_011516963.2 | 1 | 92 | XP_011515265.1 | |
| AK022122.1 | 1 | 79 | BAB13965.1 | |
| AK300762.1 | 1 | 71 | ||
| BC013136.1 | 1 | 67 | AAH13136.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470235 | Chr.8:120424982 - 120514507 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map