Assay Details
Target Gene Details
Entrez Gene ID: | 25897 |
Gene Name: | ring finger protein 19A, RBR E3 ubiquitin protein ligase |
Gene Aliases: |
RNF19 |
Location: |
Chr.8:100257059-100336218 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF19A | NM_001280539.1 | 11 | 2225 | NP_001267468.1 |
| NM_015435.4 | 11 | 2256 | NP_056250.3 | |
| NM_183419.3 | 10 | 2332 | NP_904355.1 | |
| XM_005250853.3 | 10 | 2294 | XP_005250910.1 | |
| XM_011516956.2 | 9 | 2155 | XP_011515258.1 | |
| XM_017013302.1 | 11 | 2417 | XP_016868791.1 | |
| XM_017013303.1 | 11 | 2327 | XP_016868792.1 | |
| XM_017013304.1 | 9 | 2112 | XP_016868793.1 | |
| XM_017013305.1 | 8 | 1812 | XP_016868794.1 | |
| AB029316.1 | 11 | 2256 | BAB39353.1 | |
| AJ242975.1 | 5 | 677 | CAB45132.1 | |
| AK027070.1 | 5 | 675 | BAB15647.1 | |
| AL122096.1 | 3 | 372 | CAB59264.1 | |
| BC093938.1 | 9 | 2028 | AAH93938.1 | |
| BC093940.2 | 9 | 2028 | AAH93940.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv820040 | Chr.8:100257101 - 100259800 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map