Assay Details
Target Gene Details
Entrez Gene ID: | 440275 |
Gene Name: | eukaryotic translation initiation factor 2 alpha kinase 4 |
Gene Aliases: |
GCN2, PVOD2 |
Location: |
Chr.15:39934101-40035596 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2AK4 | NM_001013703.3 | 6 | 767 | NP_001013725.2 |
| XM_005254392.2 | 6 | 790 | XP_005254449.1 | |
| XM_011521599.2 | 6 | 790 | XP_011519901.1 | |
| XM_011521600.2 | 6 | 790 | XP_011519902.1 | |
| XM_017022219.1 | 6 | 790 | XP_016877708.1 | |
| AB037759.1 | 2 | 233 | BAA92576.1 | |
| AL832907.1 | 1 | 111 | CAH10626.1 | |
| BC078179.1 | 6 | 745 | AAH78179.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581639 | Chr.15:39912419 - 39972860 on Build GRCh38 | Loss |
|
| nsv832983 | Chr.15:39806565 - 39956638 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map