Assay Details
Target Gene Details
Entrez Gene ID: | 3163 |
Gene Name: | heme oxygenase 2 |
Gene Aliases: |
HO-2 |
Location: |
Chr.16:4474697-4510347 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMOX2 | NM_001127206.2 | 1 | 49 | NP_001120678.1 |
| XM_017023197.1 | XP_016878686.1 | |||
| BE561721.1 | 1 | 86 | ||
| BI225653.1 | 1 | 50 |
Target Gene Details
Entrez Gene ID: | 57407 |
Gene Name: | NmrA-like family domain containing 1 |
Gene Aliases: |
HSCARG, SDR48A1 |
Location: |
Chr.16:4461677-4476320 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NMRAL1 | NM_020677.4 | 1 | 145 | NP_065728.1 |
| XM_006720905.3 | 2 | 126 | XP_006720968.1 | |
| XM_011522567.2 | XP_011520869.1 | |||
| XM_017023488.1 | XP_016878977.1 | |||
| XM_017023492.1 | XP_016878981.1 | |||
| AF225419.1 | 1 | 150 | AAG09721.1 | |
| BC007364.2 | AAH07364.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160332 | Chr.16:4367643 - 4486554 on Build GRCh38 | Duplication |
|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
| esv23732 | Chr.16:4340813 - 4476244 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map