Assay Details
Target Gene Details
Entrez Gene ID: | 4124 |
Gene Name: | mannosidase alpha class 2A member 1 |
Gene Aliases: |
AMan II, GOLIM7, MANA2, MANII |
Location: |
Chr.5:109689366-109869625 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAN2A1 | NM_002372.3 | 1 | 247 | NP_002363.2 |
| BX341412.2 | 1 | 158 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1025801 | Chr.5:109627949 - 109731218 on Build GRCh38 | Gain |
|
| nsv1144116 | Chr.5:109689399 - 109689799 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map