Assay Details
Target Gene Details
Entrez Gene ID: | 83875 |
Gene Name: | beta-carotene oxygenase 2 |
Gene Aliases: |
B-DIOX-II, BCDO2 |
Location: |
Chr.11:112175485-112218929 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCO2 | NM_001037290.2 | 12 | 1685 | NP_001032367.2 |
| NM_001256397.1 | 12 | 1667 | NP_001243326.1 | |
| NM_001256398.1 | 10 | 1578 | NP_001243327.1 | |
| NM_001256400.1 | 11 | 1480 | NP_001243329.1 | |
| NM_031938.5 | 12 | 1797 | NP_114144.4 | |
| AB096252.1 | 12 | 1651 | ||
| AF276432.1 | 12 | 2294 | AAK69433.1 | |
| AJ290393.1 | 12 | 1647 | CAC27994.1 | |
| AK027801.1 | 12 | 1662 | BAB55379.1 | |
| AK297853.1 | 11 | 1471 | ||
| AK315300.1 | 12 | 1789 | ||
| BC041656.1 | 10 | 1541 |
More Information
Set Membership: |
|

Genomic Map