Assay Details
Target Gene Details
Entrez Gene ID: | 2897 |
Gene Name: | glutamate ionotropic receptor kainate type subunit 1 |
Gene Aliases: |
EAA3, EEA3, GLR5, GLUR5, GluK1, gluR-5 |
Location: |
Chr.21:29536933-29940052 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIK1 | NM_000830.4 | NP_000821.1 | ||
| NM_001320616.1 | NP_001307545.1 | |||
| NM_001320618.1 | NP_001307547.1 | |||
| NM_001320621.1 | NP_001307550.1 | |||
| NM_175611.2 | NP_783300.1 | |||
| XM_005260942.1 | XP_005260999.1 | |||
| XM_005260944.3 | XP_005261001.1 | |||
| XM_006723991.1 | XP_006724054.1 | |||
| AJ249208.1 | CAC80546.1 | |||
| AK294353.1 | ||||
| AK296404.1 | ||||
| L19058.1 | AAA52568.1 | |||
| U16125.1 | AAA95961.1 |
Target Gene Details
Entrez Gene ID: | 100379661 |
Gene Name: | GRIK1 antisense RNA 2 |
Gene Aliases: |
C21orf41, NCRNA00258 |
Location: |
Chr.21:29596040-29630747 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIK1-AS2 | NR_033368.1 | 1 | 532 | |
| AY081145.1 | 1 | 534 | AAL91139.1 |
More Information
Set Membership: |
|

Genomic Map