Assay Details
Target Gene Details
Entrez Gene ID: | 2705 |
Gene Name: | gap junction protein beta 1 |
Gene Aliases: |
CMTX, CMTX1, CX32 |
Location: |
Chr.X:71215212-71225215 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJB1 | NM_000166.5 | 2 | 1240 | NP_000157.1 |
| NM_001097642.2 | 2 | 1189 | NP_001091111.1 | |
| XM_011530907.2 | 2 | 1288 | XP_011529209.1 | |
| XM_017029408.1 | 3 | 1409 | XP_016884897.1 | |
| BC002805.1 | 2 | 1149 | AAH02805.1 | |
| BC022426.1 | 2 | 1220 | AAH22426.1 | |
| BC039198.1 | 2 | 1162 | AAH39198.1 | |
| X04325.1 | 2 | 1156 | CAA27856.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508781 | Chr.X:71205875 - 71286242 on Build GRCh38 | Insertion |
|
| esv33082 | Chr.X:70420685 - 71888183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map