Assay Details
Target Gene Details
Entrez Gene ID: | 991 |
Gene Name: | cell division cycle 20 |
Gene Aliases: |
CDC20A, bA276H19.3, p55CDC |
Location: |
Chr.1:43358955-43363203 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC20 | NM_001255.2 | NP_001246.2 | ||
| AF099644.1 | AAD16405.1 | |||
| AK225504.1 | ||||
| AK302539.1 | ||||
| AK303844.1 | ||||
| AK312780.1 | ||||
| BC000624.2 | AAH00624.1 | |||
| BC001088.2 | AAH01088.1 | |||
| BC006272.2 | AAH06272.1 | |||
| BC009425.1 | AAH09425.1 | |||
| BC009426.1 | AAH09426.1 | |||
| BC010044.2 | AAH10044.1 | |||
| BC012803.2 | AAH12803.1 | |||
| BC012827.2 | AAH12827.1 | |||
| BC013303.2 | AAH13303.1 | |||
| BC015998.1 | AAH15998.1 | |||
| BC024257.1 | AAH24257.1 | |||
| BC031294.1 | AAH31294.1 | |||
| BC110321.1 | AAI10322.1 | |||
| BT007388.1 | AAP36052.1 | |||
| U05340.1 | AAA19017.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546128 | Chr.1:43301467 - 43451738 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map