Assay Details
Target Gene Details
Entrez Gene ID: | 79036 |
Gene Name: | KxDL motif containing 1 |
Gene Aliases: |
BORCS4, C19orf50, KXDL, MST096, MSTP096 |
Location: |
Chr.19:18557762-18569387 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KXD1 | NM_001171948.1 | NP_001165419.1 | ||
| NM_001171949.1 | NP_001165420.1 | |||
| NM_024069.3 | NP_076974.2 | |||
| XM_006722883.2 | XP_006722946.1 | |||
| XM_011528272.2 | XP_011526574.1 | |||
| XM_011528273.2 | XP_011526575.1 | |||
| XM_017027281.1 | XP_016882770.1 | |||
| AF173381.1 | AAQ13622.1 | |||
| AK091145.1 | ||||
| AK098346.1 | ||||
| BC001080.2 | AAH01080.1 | |||
| BC006479.1 | AAH06479.1 | |||
| BM781878.1 | ||||
| BX648739.1 | ||||
| DA542130.1 | ||||
| DA616317.1 |
Target Gene Details
Entrez Gene ID: | 7311 |
Gene Name: | ubiquitin A-52 residue ribosomal protein fusion product 1 |
Gene Aliases: |
CEP52, HUBCEP52, L40, RPL40 |
Location: |
Chr.19:18563766-18577460 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBA52 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833776 | Chr.19:18527218 - 18671454 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map