Assay Details
Target Gene Details
Entrez Gene ID: | 10946 |
Gene Name: | splicing factor 3a subunit 3 |
Gene Aliases: |
PRP9, PRPF9, SAP61, SF3a60 |
Location: |
Chr.1:37956975-37990110 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SF3A3 | NM_001320830.1 | NP_001307759.1 | ||
| NM_006802.3 | NP_006793.1 | |||
| AK128438.1 | ||||
| AK222554.1 | BAD96274.1 | |||
| AK291181.1 | ||||
| AK292116.1 | ||||
| AK301424.1 | ||||
| BC002395.2 | AAH02395.1 | |||
| BC011523.1 | AAH11523.1 | |||
| BM455074.1 | ||||
| BM905122.1 | ||||
| N91991.1 | ||||
| U08815.1 | AAA19625.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008462 | Chr.1:37938366 - 37982665 on Build GRCh38 | Gain |
|
| nsv945885 | Chr.1:37957293 - 37958291 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map