Assay Details
Target Gene Details
Entrez Gene ID: | 23536 |
Gene Name: | adenosine deaminase, tRNA specific 1 |
Gene Aliases: |
HADAT1 |
Location: |
Chr.16:75596879-75623323 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAT1 | NM_001324444.1 | NP_001311373.1 | ||
| NM_001324445.1 | NP_001311374.1 | |||
| NM_001324446.1 | NP_001311375.1 | |||
| NM_001324448.1 | NP_001311377.1 | |||
| NM_001324449.1 | NP_001311378.1 | |||
| NM_001324450.1 | NP_001311379.1 | |||
| NM_001324451.1 | NP_001311380.1 | |||
| NM_001324452.1 | NP_001311381.1 | |||
| NM_001324453.1 | NP_001311382.1 | |||
| NM_012091.4 | NP_036223.2 | |||
| AF125188.1 | AAD48376.1 | |||
| AK001687.1 | BAA91837.1 | |||
| AK296834.1 | ||||
| AK314652.1 | ||||
| BC002758.2 | AAH02758.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3639101 | Chr.16:75599180 - 75622670 on Build GRCh38 | Loss |
|
| nsv1065543 | Chr.16:75597408 - 75766272 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map