Assay Details
Target Gene Details
Entrez Gene ID: | 2200 |
Gene Name: | fibrillin 1 |
Gene Aliases: |
ACMICD, ECTOL1, FBN, GPHYSD2, MASS, MFLS, MFS1, OCTD, SGS, SSKS, WMS, WMS2 |
Location: |
Chr.15:48408306-48645788 on Build GRCh38 |
Assay Gene Location: | Within Exon 66 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBN1 | NM_000138.4 | 66 | 11439 | NP_000129.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv817692 | Chr.15:48402014 - 48409031 on Build GRCh38 | Loss |
|
| esv3636415 | Chr.15:48397504 - 48409161 on Build GRCh38 | Loss |
|
| nsv1047597 | Chr.15:48345836 - 48456074 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map